Anti-CHEK1 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 2G1D5]

Leverandør: Novus Biologicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

NOVUNB100-1768FEA 4580 DKK
NOVUNB100-1768F
Anti-CHEK1 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 2G1D5]
Antistoffer
Type: Primary
Antigen: CHEK1 (checkpoint kinase 1)
Clonality: monoclonal
Clone: 2G1D5
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR