Anti-DNA Polymerase iota Rabbit Polyclonal Antibody

Leverandør: Novus Biologicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

NOVUNB100-175EA 3590 DKK
NOVUNB100-175 NOVUNB100-175SS
Anti-DNA Polymerase iota Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: DNA Polymerase iota
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR