Anti-PECAM1 Rat Monoclonal Antibody (FITC (Fluorescein)) [clone: MEC 7.46]

Leverandør: Novus Biologicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

NOVUNB100-1642FEA 4980 DKK
NOVUNB100-1642F
Anti-PECAM1 Rat Monoclonal Antibody (FITC (Fluorescein)) [clone: MEC 7.46]
Antistoffer
Type: Primary
Antigen: PECAM1 (platelet/endothelial cell adhesion molecule 1)
Clonality: monoclonal
Clone: MEC 7.46
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR