Anti-HIF1A Mouse Monoclonal Antibody (Biotin) [clone: ESEE122]

Leverandør: Novus Biologicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

NOVUNB100-131BEA 5520 DKK
NOVUNB100-131B
Anti-HIF1A Mouse Monoclonal Antibody (Biotin) [clone: ESEE122]
Antistoffer
Type: Primary
Antigen: HIF1A (hypoxia inducible factor 1, alpha subunit (basic helix-loop-helix transcription factor))
Clonality: monoclonal
Clone: ESEE122
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR