Anti-HIF-1 beta Mouse Monoclonal Antibody (DyLight® 650) [clone: H1beta234]

Leverandør: Novus Biologicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

NOVUNB100-124CEA 5520 DKK
NOVUNB100-124C
Anti-HIF-1 beta Mouse Monoclonal Antibody (DyLight® 650) [clone: H1beta234]
Antistoffer
Type: Primary
Antigen: HIF-1 beta
Clonality: monoclonal
Clone: H1beta234
Conjugation: DyLight® 650
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR