Anti-HIF1A Mouse Monoclonal Antibody [clone: H1alpha67] (DyLight® 550)

Leverandør: Novus Biologicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

NOVUNB100-123REA 5930 DKK
NOVUNB100-123R
Anti-HIF1A Mouse Monoclonal Antibody [clone: H1alpha67] (DyLight® 550)
Antistoffer
Mouse Monoclonal HIF-1 alpha Antibody (H1alpha67) [DyLight 550]. Tested Applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin. Tested Reactivity: Human, Mouse, Rat, Bovine, Ferret, Primate, Porcine, Sheep.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:500-1:1000

Type: Primary
Antigen: HIF1A
Clonality: Monoclonal
Clone:
Conjugation: DyLight® 550
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR