Anti-HIF1A Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: H1alpha67]

Leverandør: Novus Biologicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

NOVUNB100-123FEA 5260 DKK
NOVUNB100-123F
Anti-HIF1A Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: H1alpha67]
Antistoffer
Type: Primary
Antigen: HIF1A (hypoxia inducible factor 1, alpha subunit (basic helix-loop-helix transcription factor))
Clonality: monoclonal
Clone: H1alpha67
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR