Anti-DNA Polymerase epsilon Mouse Monoclonal Antibody (DyLight® 680) [clone: 3C5.1]
Leverandør: Novus Biologicals
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
NOVUNB100-115FREA 4590 DKK
NOVUNB100-115FR
Anti-DNA Polymerase epsilon Mouse Monoclonal Antibody (DyLight® 680) [clone: 3C5.1]
Antistoffer
Type: Primary
Antigen: DNA Polymerase epsilon
Clonality: monoclonal
Clone: 3C5.1
Conjugation: DyLight® 680
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Bestil nu