Anti-OGG1 Rabbit Polyclonal Antibody (FITC (Fluorescein))

Leverandør: Novus Biologicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

NOVUNB100-106FEA 5520 DKK
NOVUNB100-106F
Anti-OGG1 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Antistoffer
Type: Primary
Antigen: OGG1 (8-oxoguanine DNA glycosylase)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR