Anti-APEX1 Rabbit Polyclonal Antibody
Leverandør: Novus Biologicals
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
NOVUNB100-101SSEA 1870 DKK
NOVUNB100-101SS NOVUNB100-101
Anti-APEX1 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: APEX1 (APEX nuclease (multifunctional DNA repair enzyme) 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Bestil nu