Anti-E2F3 Rabbit Polyclonal Antibody

Leverandør: Boster
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BSBTPA2142EA 4110 DKK
BSBTPA2142
Anti-E2F3 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: E2F3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR