Anti-NADPH oxidase 4 Rabbit Polyclonal Antibody

Leverandør: Boster
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BSBTPA1929EA 4110 DKK
BSBTPA1929
Anti-NADPH oxidase 4 Rabbit Polyclonal Antibody
Antistoffer
Rabbit IgG polyclonal antibody for NADPH oxidase 4 (NOX4) detection. Tested with WB, IHC-P in Human; Mouse; Rat.

Reconstitute with distilled water.

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: NADPH oxidase 4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR