Anti-Cpn10 Rabbit Polyclonal Antibody
Leverandør: Boster
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
BSBTPA1790EA 4110 DKK
BSBTPA1790
Anti-Cpn10 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: Cpn10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat