Anti-IL3 Rabbit Polyclonal Antibody
Leverandør: Boster
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
BSBTPA1728EA 4110 DKK
BSBTPA1728
Anti-IL3 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: IL3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse