Anti-Hsp105 Rabbit Polyclonal Antibody

Leverandør: Boster
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BSBTPA1608EA 4110 DKK
BSBTPA1608
Anti-Hsp105 Rabbit Polyclonal Antibody
Antistoffer
Rabbit IgG polyclonal antibody for Heat shock protein 105 kDa(HSPH1) detection. Tested with WB, IHC-P, IHC-F in Human; Mouse; Rat.

Reconstitute with distilled water.

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: Hsp105
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR