Anti-HSD17B2 Rabbit Polyclonal Antibody

Leverandør: Boster
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BSBTPA1604EA 4110 DKK
BSBTPA1604
Anti-HSD17B2 Rabbit Polyclonal Antibody
Antistoffer
Rabbit IgG polyclonal antibody for Estradiol 17-beta-dehydrogenase 2(HSD17B2) detection. Tested with WB, IHC-P, IHC-F in Human.

Reconstitute with distilled water.

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: HSD17B2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR