Anti-CaMKK Rabbit Polyclonal Antibody
Leverandør: Boster
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
BSBTPA1520EA 4110 DKK
BSBTPA1520
Anti-CaMKK Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: CaMKK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the N-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat