Anti-intestinal FABP Rabbit Polyclonal Antibody
Leverandør: Boster
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
BSBTPA1452EA 4110 DKK
BSBTPA1452
Anti-intestinal FABP Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: intestinal FABP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the N-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat