Anti-Myelin Basic Protein Rabbit Polyclonal Antibody

Leverandør: Boster
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BSBTPA1050EA 4110 DKK
BSBTPA1050
Anti-Myelin Basic Protein Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: Myelin Basic Protein
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR