Anti-CD40 Rabbit Polyclonal Antibody

Leverandør: Boster
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BSBTPA1019-1EA 4110 DKK
BSBTPA1019-1
Anti-CD40 Rabbit Polyclonal Antibody
Antistoffer
Rabbit IgG polyclonal antibody for Tumor necrosis factor receptor superfamily member 5 (CD40) detection. Tested with WB in Human.

  • Reconstitute with distilled water

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: CD40
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the N-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR