Anti-Annexin V Rabbit Polyclonal Antibody

Leverandør: Boster
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BSBTPA1008EA 4110 DKK
BSBTPA1008
Anti-Annexin V Rabbit Polyclonal Antibody
Antistoffer
Rabbit IgG polyclonal antibody for Annexin A5 (ANXA5) detection. Tested with WB, IHC-P, ICC in Human;Mouse;Rat.

Reconstitute with distilled water.

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: Annexin V
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR