Anti-Caldesmon (smooth) Mouse Monoclonal Antibody [clone: CD-7]

Leverandør: Boster
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BSBTMA1009EA 4110 DKK
BSBTMA1009
Anti-Caldesmon (smooth) Mouse Monoclonal Antibody [clone: CD-7]
Antistoffer
Mouse IgG monoclonal antibody for Caldesmon (smooth), caldesmon 1 (CALD1) detection. Tested with WB, IHC-F in Human;mouse;rat. No cross reactivity with other proteins.

Caldesmon is a potential actomyosin regulatory protein found in smooth muscle and nonmuscle cells. The predicted smooth-muscle polypeptide is 793 amino acids long. The high molecular weight caldesmon(h-CaD) is predominantly expressed in smooth muscles, whereas the low molecular weight caldesmon(l-CaD) is widely distributed in nonmuscle tissues and cells. Hayashi et al.(1992) demonstrated that the human CDM gene is composed of 14 exons.

Mouse ascites fluid, 1.2% sodium acetate, 2mg BSA, with 0.01mg NaN3 as preservative.

Other applications have not been tested.
Optimal dilutions should be determined by end users.

Type: Primary
Antigen: Caldesmon (smooth)
Clonality: monoclonal
Clone: CD-7
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rabbit
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR