Anti-IgG Goat Polyclonal Antibody (CF™ 770)
Leverandør: Biotium
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
20288-1EA 1080 DKK
20288-1 20288.
Anti-IgG Goat Polyclonal Antibody (CF™ 770)
Antistoffer
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF® 770 Dye
Epitope:
Host: Goat
Isotype: none
Reactivity: Human