Anti-IgG Goat Polyclonal Antibody (CF™ 647)
Leverandør: Biotium
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
20282-1EA 1080 DKK
20282-1 20282.
Anti-IgG Goat Polyclonal Antibody (CF™ 647)
Antistoffer
Type: Secondary
Antigen: IgG (Rabbit) (rabbit IgG)
Clonality: polyclonal
Clone:
Conjugation: CF® 647 Dye
Epitope:
Host: Goat
Isotype: none
Reactivity: Rabbit