Anti-IgG2b Goat Polyclonal Antibody (CF™ 488A)

Leverandør: Biotium
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

20266-1EA 1150 DKK
20266-1 20266.
Anti-IgG2b Goat Polyclonal Antibody (CF™ 488A)
Antistoffer
Type: Secondary
Antigen: IgG2b (Mouse) (mouse IgG2b)
Clonality: polyclonal
Clone:
Conjugation: CF® 488A Dye
Epitope:
Host: Goat
Isotype: none
Reactivity: Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR