Anti-IgG2b Goat Polyclonal Antibody (CF™ 350)
Leverandør: Biotium
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
20265.EA 2780 DKK
20265. 20265-1
Anti-IgG2b Goat Polyclonal Antibody (CF™ 350)
Antistoffer
Type: Secondary
Antigen: IgG2b (Mouse) (mouse IgG2b)
Clonality: polyclonal
Clone:
Conjugation: CF® 350 Dye
Epitope:
Host: Goat
Isotype: none
Reactivity: Mouse