Anti-IgG Donkey Polyclonal Antibody (CF™ 770)

Leverandør: Biotium
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

20242.EA 2520 DKK
20242. 20242-1
Anti-IgG Donkey Polyclonal Antibody (CF™ 770)
Antistoffer
Type: Secondary
Antigen: IgG (Guinea pig) (guinea pig IgG)
Clonality: polyclonal
Clone:
Conjugation: CF® 770 Dye
Epitope:
Host: Donkey
Isotype: none
Reactivity: Guinea pig
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR