Anti-IgG Rabbit Polyclonal Antibody (CF™ 640R)

Leverandør: Biotium
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

20201.EA 2170 DKK
20201. 20201-1
Anti-IgG Rabbit Polyclonal Antibody (CF™ 640R)
Antistoffer
Type: Secondary
Antigen: IgG (Rat) (rat IgG)
Clonality: polyclonal
Clone:
Conjugation: CF® 640R Dye
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR