Anti-IgG Goat Polyclonal Antibody (CF™ 594)

Leverandør: Biotium
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

20118-1EA 819 DKK
20118-1 20118.
Anti-IgG Goat Polyclonal Antibody (CF™ 594)
Antistoffer
Type: Secondary
Antigen: IgG (Guinea pig) (guinea pig IgG)
Clonality: polyclonal
Clone:
Conjugation: CF® 594 Dye
Epitope:
Host: Goat
Isotype: none
Reactivity: Guinea pig
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR