Anti-IgG Rabbit Polyclonal Antibody (CF™ 488A)

Leverandør: Biotium
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

20071-1EA 1130 DKK
20071-1 20071.
Anti-IgG Rabbit Polyclonal Antibody (CF™ 488A)
Antistoffer
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF® 488A Dye
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR