Anti-IgG Goat Polyclonal Antibody (CF™ 680)
Leverandør: Biotium
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
20065.EA 2510 DKK
20065. 20065-1
Anti-IgG Goat Polyclonal Antibody (CF™ 680)
Antistoffer
Type: Secondary
Antigen: IgG (Mouse) (mouse IgG)
Clonality: polyclonal
Clone:
Conjugation: CF® 680 Dye
Epitope:
Host: Goat
Isotype: none
Reactivity: Mouse