Anti-IgG Donkey Polyclonal Antibody (CF™ 680)

Leverandør: Biotium
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering


20062.EA 2510 DKK
20062. 20062-1
Anti-IgG Donkey Polyclonal Antibody (CF™ 680)
Antistoffer
Type: Secondary
Antigen: IgG (Sheep) (sheep IgG)
Clonality: polyclonal
Clone:
Conjugation: CF® 680 Dye
Epitope:
Host: Donkey
Isotype: none
Reactivity: Sheep
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR