Anti-IgG Rabbit Polyclonal Antibody (CF™ 488A)
Leverandør: Biotium
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
20025.EA 2450 DKK
20025. 20025-1
Anti-IgG Rabbit Polyclonal Antibody (CF™ 488A)
Antistoffer
Type: Secondary
Antigen: IgG (Rat) (rat IgG)
Clonality: polyclonal
Clone:
Conjugation: CF® 488A Dye
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Rat