Anti-IgG Donkey Polyclonal Antibody (CF™ 488A)
Leverandør: Biotium
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
20024-1EA 1020 DKK
20024-1 20024.
Anti-IgG Donkey Polyclonal Antibody (CF™ 488A)
Antistoffer
Type: Secondary
Antigen: IgG (Sheep) (sheep IgG)
Clonality: polyclonal
Clone:
Conjugation: CF® 488A Dye
Epitope:
Host: Donkey
Isotype: none
Reactivity: Sheep