Anti-GPR32 Rabbit Polyclonal Antibody

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB150642-50EA 6450 DKK
ABCAAB150642-50
Anti-GPR32 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: GPR32 (G protein-coupled receptor 32)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: extracellular domain
Host: Rabbit
Isotype: none
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR