Anti-GPR18 Rabbit Polyclonal Antibody

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB150618-50EA 6230 DKK
ABCAAB150618-50
Anti-GPR18 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: GPR18 (G protein-coupled receptor 18)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: cytoplasmic domain
Host: Rabbit
Isotype: none
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR