Anti-KMT6 / EZH2 Rabbit Monoclonal Antibody
Leverandør: Abcam
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ABCAAB150433-100EA 7060 DKK
ABCAAB150433-100
Anti-KMT6 / EZH2 Rabbit Monoclonal Antibody
Antistoffer
Type: Primary
Antigen: KMT6 / EZH2
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestil nu