Anti-CD1B Rabbit Monoclonal Antibody
Leverandør: Abcam
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ABCAAB150410-100EA 6430 DKK
ABCAAB150410-100
Anti-CD1B Rabbit Monoclonal Antibody
Antistoffer
Type: Primary
Antigen: CD1B
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestil nu