Anti-MAP4K4 Rabbit Monoclonal Antibody
Leverandør: Abcam
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ABCAAB150300-100EA 6540 DKK
ABCAAB150300-100
Anti-MAP4K4 Rabbit Monoclonal Antibody
Antistoffer
Type: Primary
Antigen: MAP4K4
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestil nu