Anti-VHL Mouse Monoclonal Antibody [clone: 1E1]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB140989-100EA 5280 DKK
ABCAAB140989-100
Anti-VHL Mouse Monoclonal Antibody [clone: 1E1]
Type: Primary
Antigen: VHL (von Hippel-Lindau tumor suppressor, E3 ubiquitin protein ligase)
Clonality: monoclonal
Clone: 10
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR