Anti-PDCD1 Mouse Monoclonal Antibody [clone: J116]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB140950-50EA 6490 DKK
ABCAAB140950-50
Anti-PDCD1 Mouse Monoclonal Antibody [clone: J116]
Antistoffer
Type: Primary
Antigen: PDCD1 (programmed cell death 1)
Clonality: monoclonal
Clone: J116
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR