Anti-OR51E2 Rabbit Polyclonal Antibody

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB140907-50EA 6290 DKK
ABCAAB140907-50
Anti-OR51E2 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: OR51E2 (olfactory receptor, family 51, subfamily E, member 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR