Anti-Ghrelin O acyltransferase Rabbit Polyclonal Antibody
Leverandør: Abcam
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ABCAAB140889-50EA 6480 DKK
ABCAAB140889-50
Anti-Ghrelin O acyltransferase Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: Ghrelin O acyltransferase
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human