Anti-P2RY11 Rabbit Polyclonal Antibody

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB140878-50EA 4100.24 DKK
ABCAAB140878-50
Anti-P2RY11 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: P2RY11 (purinergic receptor P2Y, G-protein coupled, 11)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: extracellular domain
Host: Rabbit
Isotype: none
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR