Anti-FCGR1A Mouse Monoclonal Antibody [clone: 3D3]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB140779-100EA 3688.6 DKK
ABCAAB140779-100
Anti-FCGR1A Mouse Monoclonal Antibody [clone: 3D3]
Antistoffer
Type: Primary
Antigen: FCGR1A (Fc fragment of IgG, high affinity Ia, receptor (CD64))
Clonality: monoclonal
Clone: 3D3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR