Anti-ADRA2C Rabbit Polyclonal Antibody
Leverandør: Abcam
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ABCAAB140702-50EA 6480 DKK
ABCAAB140702-50
Anti-ADRA2C Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: ADRA2C (adrenoceptor alpha 2C)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human