Anti-PSMD1 Rabbit Polyclonal Antibody

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB140682-100EA 6620 DKK
ABCAAB140682-100
Anti-PSMD1 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: PSMD1 (proteasome (prosome, macropain) 26S subunit, non-ATPase, 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR