Anti-SRY Mouse Monoclonal Antibody [clone: 3C8]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB140309-100EA 4098.45 DKK
ABCAAB140309-100
Anti-SRY Mouse Monoclonal Antibody [clone: 3C8]
Type: Primary
Antigen: SRY (sex determining region Y)
Clonality: monoclonal
Clone: 3C8
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR