Anti-CD82 Mouse Monoclonal Antibody [clone: C33]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB140238-100EA 6450 DKK
ABCAAB140238-100
Anti-CD82 Mouse Monoclonal Antibody [clone: C33]
Antistoffer
Type: Primary
Antigen: CD82 (CD82 molecule)
Clonality: monoclonal
Clone: C33
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR