Anti-CARD14 Rabbit Polyclonal Antibody

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB140203-100EA 6450 DKK
ABCAAB140203-100
Anti-CARD14 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: CARD14 (caspase recruitment domain family, member 14)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR